missing translation for 'onlineSavingsMsg'
Learn More

Phenylsilane 97.0+%, TCI America™

Catalog No. p-7135057
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
P12915ML 5 mL
P129125ML 25 mL
2 options

Catalog No. P12915ML

Supplier: TCI America P12915ML

Only null left

Chemical Identifiers

CAS 694-53-1
Molecular Formula C6H5Si
Molecular Weight (g/mol) 105.191
MDL Number MFCD00013478
InChI Key XJWOWXZSFTXJEX-UHFFFAOYSA-N
Synonym phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
PubChem CID 6327628
IUPAC Name phenylsilicon
SMILES C1=CC=C(C=C1)[Si]

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 mL
UN Number 1993
Formula Weight 108.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenylsilane
RTECSNumber VV4825000
TSCA Yes