missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl bromide, 98%

CAS: 34837-55-3 | C6H5BrSe | 235.981 g/mol

$42.73 - $443.10

Chemical Identifiers

CAS 34837-55-3
Molecular Formula C6H5BrSe
Molecular Weight (g/mol) 235.981
MDL Number MFCD00000047
InChI Key LCEFEIBEOBPPSJ-UHFFFAOYSA-N
Synonym phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
PubChem CID 123446
IUPAC Name phenyl selenohypobromite
SMILES C1=CC=C(C=C1)[Se]Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1190603
View Documents
Thermo Scientific Chemicals
A1190603
1 g
Each for $42.73
Only null left
 
AAA1190606
View Documents
Thermo Scientific Chemicals
A1190606
5 g
Each for $139.29
Only null left
 
AAA1190614
View Documents
Thermo Scientific Chemicals
A1190614
25 g
Each for $443.10
Only null left
 
Description

Description

Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.

Solubility
Soluble in toluene, very faint turbidity.

Notes
Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

34837-55-3
235.981
LCEFEIBEOBPPSJ-UHFFFAOYSA-N
123446
C1=CC=C(C=C1)[Se]Br
C6H5BrSe
MFCD00000047
phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
phenyl selenohypobromite

Specifications

34837-55-3
107°C to 108°C (15 mmHg)
MFCD00000047
UN2928
Moisture sensitive
Soluble in toluene,very faint turbidity.
C1=CC=C(C=C1)[Se]Br
235.981
235.98
Phenylselenenyl bromide
56°C to 62°C
C6H5BrSe
1 g
2039772
phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
LCEFEIBEOBPPSJ-UHFFFAOYSA-N
phenyl selenohypobromite
123446
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H331-H314-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 252-238-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only