missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl bromide, 98%

CAS: 34837-55-3 | C6H5BrSe | 235.981 g/mol

Supplier:  Thermo Scientific Chemicals A1190606

 View more versions of this product

Catalog No. AAA1190606


Only null left
Explore available promotions

Description

Description

Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.

Solubility
Soluble in toluene, very faint turbidity.

Notes
Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Phenylselenenyl bromide
56°C to 62°C
C6H5BrSe
5 g
2039772
phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
LCEFEIBEOBPPSJ-UHFFFAOYSA-N
phenyl selenohypobromite
123446
98%
34837-55-3
107°C to 108°C (15 mmHg)
MFCD00000047
UN2928
Moisture sensitive
Soluble in toluene,very faint turbidity.
C1=CC=C(C=C1)[Se]Br
235.981
235.98

Chemical Identifiers

34837-55-3
235.981
LCEFEIBEOBPPSJ-UHFFFAOYSA-N
123446
C1=CC=C(C=C1)[Se]Br
C6H5BrSe
MFCD00000047
phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
phenyl selenohypobromite
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H331-H314-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 252-238-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only