missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic dichloride, 90+%

CAS: 824-72-6 | C6H5Cl2OP | 194.98 g/mol

Supplier:  Thermo Scientific Chemicals A137770B

 View more versions of this product

Catalog No. AAA137770B

Only null left
Explore available promotions

Description

Description

Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

Solubility
Miscible with benzene, chloroform, dimethyl sulfoxide and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and water.
Specifications

Specifications

Phenylphosphonic dichloride
1°C to 3°C
258°C
C6H5Cl2OP
MFCD00002070
UN3265
Moisture sensitive
Miscible with benzene,chloroform,dimethyl sulfoxide and carbon tetrachloride.
ClP(Cl)(=O)C1=CC=CC=C1
194.98
194.99
824-72-6
1.379
204°C (399°F)
1.56
1000 g
1072240
phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
IBDMRHDXAQZJAP-UHFFFAOYSA-N
dichlorophosphorylbenzene
69990
≥90%

Chemical Identifiers

824-72-6
194.98
IBDMRHDXAQZJAP-UHFFFAOYSA-N
69990
ClP(Cl)(=O)C1=CC=CC=C1
C6H5Cl2OP
MFCD00002070
phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
dichlorophosphorylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 212-534-3

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only