missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic dichloride, 90+%

Catalog No. p-7021445
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1377718 50 g
AAA1377730 250 g
AAA137770B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1377718 Supplier Thermo Scientific Chemicals Supplier No. A1377718

CAS: 824-72-6 | C6H5Cl2OP | 194.98 g/mol

Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

Solubility
Miscible with benzene, chloroform, dimethyl sulfoxide and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and water.

Chemical Identifiers

CAS 824-72-6
Molecular Formula C6H5Cl2OP
Molecular Weight (g/mol) 194.98
MDL Number MFCD00002070
InChI Key IBDMRHDXAQZJAP-UHFFFAOYSA-N
Synonym phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
PubChem CID 69990
IUPAC Name dichlorophosphorylbenzene
SMILES ClP(Cl)(=O)C1=CC=CC=C1

Specifications

Melting Point 1°C to 3°C
Density 1.379
Boiling Point 258°C
Flash Point 204°C (399°F)
Refractive Index 1.56
Quantity 50 g
UN Number UN3265
Beilstein 1072240
Sensitivity Moisture sensitive
Solubility Information Miscible with benzene,chloroform,dimethyl sulfoxide and carbon tetrachloride.
Formula Weight 194.99
Percent Purity ≥90%
Chemical Name or Material Phenylphosphonic dichloride
Show More Show Less
Hazard Category H290-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-534-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only