missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 296680025

Catalog No. AC296680025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Phenylhydrazine
1.0900g/mL
89°C
97%
C6H8N2
C6H5NHNH2
MFCD00007573
01,838; 02,322; 06,457; 07,284
1.09
Solubility in water: 145g/L (20°C). Other solubilities: soluble in diluted acids,miscible with alcohol,ether,chloroform and,benzene,sparingly soluble in petroleum ether
C1=CC=C(C=C1)NN
108.14
CHEBI:27924
97%
100-63-0
238.0°C to 241.0°C
Authentic
Glass bottle
1.6040 to 1.6090
2.5kg
15,II,44
15,7404
hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
HKOOXMFOFWEVGF-UHFFFAOYSA-N
phenylhydrazine
7516
108.14
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause cancer.
Toxic if inhaled.
Toxic in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

EINECSNumber : 202-873-5

SDS
Documents

Documents

Promotions

Promotions