missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxylic acid, 98%

Catalog No. AAL0064814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0064814 25 g
AAL0064806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0064814 Supplier Thermo Scientific Chemicals Supplier No. L0064814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00002575
InChI Key FAQJJMHZNSSFSM-UHFFFAOYSA-N
Synonym benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
PubChem CID 11915
ChEBI CHEBI:18280
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 62°C to 67°C
Density 1.38
Boiling Point 147°C to 151°C (12 mmHg)
Quantity 25 g
Beilstein 606718
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material Phenylglyoxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-278-7
RTECSNumber DA1480000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only