missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl Alcohol, USP, Spectrum™ Chemical

P1058, 60-12-8, C8H10O

Supplier:  Spectrum Chemical Mfg Cor P1058159KGBL

 View more versions of this product

Catalog No. 18605139

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Phenylethyl Alcohol, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

60-12-8
5e-005
C8H10O
MFCD00002886
WRMNZCZEMHIOCP-UHFFFAOYSA-N
2-phenylethan-1-ol
USP
1
Steel Pail
1.531-1.534
15.9 kg
OCCC1=CC=CC=C1
122.17

Chemical Identifiers

60-12-8
122.17
WRMNZCZEMHIOCP-UHFFFAOYSA-N
OCCC1=CC=CC=C1
C8H10O
MFCD00002886
2-phenylethan-1-ol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions