missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl Alcohol, USP, Spectrum™ Chemical

Catalog No. 18605137
Change view
Click to view available options
Quantity:
100 g
1 kg
15.9 kg
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605137 100 g Amber Glass Bottle
18605140 200 L Amber Glass Bottle
18605138 1 kg Amber Glass Bottle
18605139 15.9 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605137 Supplier Spectrum Chemical Mfg Cor Supplier No. P1058100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1058, 60-12-8, C8H10O

Spectrum™ Chemical Phenylethyl Alcohol, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
IUPAC Name 2-phenylethan-1-ol
SMILES OCCC1=CC=CC=C1

Specifications

CAS Max % 1
Boiling Point 218°C (424.4°F)
Ignition Residue 5e-005
Packaging Amber Glass Bottle
Refractive Index 1.531-1.534
Quantity 100 g
Grade USP
Physical Form Viscous Liquid or Low Melting Solid