missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylethyl Alcohol, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 100 g |
| Specific Gravity | 1.017 - 1.019 |
| Percent Purity | 0.98 |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.