missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl Alcohol, 98%, Spectrum™ Chemical

Catalog No. 18612301
Change view
Click to view available options
Quantity:
100 g
1 kg
15.9 kg
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612301 100 g Amber Glass Bottle
18612302 200 L Amber Glass Bottle
18612300 1 kg Amber Glass Bottle
18612303 15.9 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612301 Supplier Spectrum Chemical Mfg Cor Supplier No. P1057100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 60-12-8

Specifications

CAS Max % 1
Boiling Point 218°C (424.4°F)
Packaging Amber Glass Bottle
Quantity 100 g
Specific Gravity 1.017 - 1.019
Molecular Weight (g/mol) 122.17
Percent Purity 0.98
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Phenylethyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.