missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015945
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015945 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015945 Supplier Merck Emd Millipore Supplier No. PHR169550MG

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 94240-17-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C9H11 NO2 · HCl
Synonym 1-(3-Hydroxyphenyl)-2-(methylamino)ethanone hydrochloride; Phenylephrone hydrochloride; 3′-Hydroxyphenyl-2-methylaminoacetophenone hydrochloride; Phenylephrone hydrochloride
Type Certified Reference Material
Quantity 50 mg
Formula Weight 201.65
Grade Certified Reference Material