missing translation for 'onlineSavingsMsg'
Learn More

Medisca Phenylephrine Hydrochloride, USP

Catalog No. 14185105
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185105 5 g
14185442 100 g
14185433 25 g
14185394 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14185105 Supplier Medisca Supplier No. 024703

Phenylephrine Hydrochloride USP crytals are white or practically white. It is odorless.

Phenylephrine Hydrochloride USP crytals are white or practically white. It is odorless.

Specifications

Chemical Name or Material Phenylephrine Hydrochloride, USP
Packaging Jar 30ml, White, HDPE, Round, 28-400, TRIVENI-DB, CT
CAS 61-76-7
Type Drug Standard
Molecular Weight (g/mol) 203.6676
Quantity 5 g
Percent Purity 98 to 102 %
Grade USP