missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014908
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014908 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014908 Supplier Merck Emd Millipore Supplier No. PHR1017500MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Phenylephrine Hydrochloride is classified under the category of sympathomimetic drugs. It is generally utilized as a nasal decongestant.

Specifications

CAS 61-76-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC6H4 CH(CH2 NHCH3)OH·HCl
MDL Number MFCD00012605
Melting Point 143°C to 145°C
Synonym (R)-(-)-Phenylephrine hydrochloride; (R)-(-)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride; (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 203.67
Grade Certified Reference Material
Show More Show Less