missing translation for 'onlineSavingsMsg'
Learn More

Phenylcyclohexane 97.0+%, TCI America™

Catalog No. p-7124469
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C049625ML 25 mL
C0496100ML 100 mL
C0496500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C049625ML Supplier TCI America Supplier No. C049625ML
Only null left

Chemical Identifiers

CAS 827-52-1
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00001451
InChI Key IGARGHRYKHJQSM-UHFFFAOYSA-N
Synonym phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
PubChem CID 13229
IUPAC Name cyclohexylbenzene
SMILES C1CCC(CC1)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 236°C
Quantity 25 mL
UN Number 3082
Formula Weight 160.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenylcyclohexane
EINECSNumber (4)-1819
RTECSNumber CZ1330000
TSCA Yes