accessibility menu, dialog, popup

UserName

CAS RN 827-52-1

IUPAC Name: cyclohexylbenzene
Synonyms: phenyl cyclohexane
Molecular Weight (g/mol): 160.26
Molecular Formula: C12H16
InChi Key: IGARGHRYKHJQSM-UHFFFAOYSA-N
SMILES: C1CCC(CC1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)