missing translation for 'onlineSavingsMsg'
Learn More

Phenylbutazone 98.0+%, TCI America™

Catalog No. P1686500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1686500G 500 g
P168625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1686500G Supplier TCI America Supplier No. P1686500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-33-9
Molecular Formula C19H20N2O2
Molecular Weight (g/mol) 308.38
MDL Number MFCD00005500
InChI Key VYMDGNCVAMGZFE-UHFFFAOYSA-N
Synonym phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion
PubChem CID 4781
ChEBI CHEBI:48574
IUPAC Name 4-butyl-1,2-diphenylpyrazolidine-3,5-dione
SMILES CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 500 g
UN Number 2811
Formula Weight 308.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylbutazone
EINECSNumber (9)-1154
RTECSNumber UQ8225000
TSCA Yes