accessibility menu, dialog, popup

UserName

CH3ONNO

CAS RN 50-33-9

IUPAC Name: 4-butyl-1,2-diphenylpyrazolidine-3,5-dione
Synonyms: phenylbutazone buzon artrizone phenylbutazone (product) diossidone diozol mephabutazon betazed azobutyl mephabutazone
Molecular Weight (g/mol): 308.38
Molecular Formula: C19H20N2O2
InChi Key: VYMDGNCVAMGZFE-UHFFFAOYSA-N
SMILES: CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)