missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Supplier: TCI America B08575G
Specifications
| Phenylboronic Acid (contains varying amounts of Anhydride) | |
| 216°C | |
| MFCD00002103 | |
| benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
| OB(O)C1=CC=CC=C1 | |
| 121.93 | |
| CHEBI:44923 | |
| Crystalline Powder |
| 98-80-6 | |
| C6H7BO2 | |
| 5 g | |
| HXITXNWTGFUOAU-UHFFFAOYSA-N | |
| phenylboronic acid | |
| 66827 | |
| 121.93 |
Chemical Identifiers
| 98-80-6 | |
| 121.93 | |
| HXITXNWTGFUOAU-UHFFFAOYSA-N | |
| 66827 | |
| phenylboronic acid |
| C6H7BO2 | |
| MFCD00002103 | |
| benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
| CHEBI:44923 |
Safety and Handling
EINECSNumber : (3)-4668
RTECSNumber : CY8575000
TSCA : Yes