missing translation for 'onlineSavingsMsg'
Learn More

Phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7118711
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08575G 5 g
B085725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08575G Supplier TCI America Supplier No. B08575G
Only null left

Chemical Identifiers

CAS 98-80-6
Molecular Formula C6H7BO2
Molecular Weight (g/mol) 121.93
MDL Number MFCD00002103
InChI Key HXITXNWTGFUOAU-UHFFFAOYSA-N
Synonym benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
PubChem CID 66827
ChEBI CHEBI:44923
IUPAC Name phenylboronic acid

Specifications

Melting Point 216°C
Quantity 5 g
SMILES OB(O)C1=CC=CC=C1
Formula Weight 121.93
Physical Form Crystalline Powder
Chemical Name or Material Phenylboronic Acid (contains varying amounts of Anhydride)
EINECSNumber (3)-4668
RTECSNumber CY8575000
TSCA Yes