missing translation for 'onlineSavingsMsg'
Learn More

Phenylazoformic acid 2-phenylhydrazide

Catalog No. AAH6033614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6033614 25 g
AAH6033606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6033614 Supplier Thermo Scientific Chemicals Supplier No. H6033614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-62-5 | C13H12N4O | 240.266 g/mol

It is a reagent for mercury determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent for mercury determination.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 538-62-5
Molecular Formula C13H12N4O
Molecular Weight (g/mol) 240.266
MDL Number MFCD00003024
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
PubChem CID 10860
IUPAC Name 1-anilino-3-phenyliminourea
SMILES C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2

Specifications

Melting Point 119°C to 123°C
Quantity 25 g
Beilstein 959475
Solubility Information Soluble in acetone. Insoluble in water.
Formula Weight 240.26
Chemical Name or Material Phenylazoformic acid 2-phenylhydrazide
EINECSNumber 208-698-0
RTECSNumber LQ9420000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only