missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide, For Dissolved Oxygen, Certified, 0.00500N ±0.00005N (0.0025M), LabChem™

$193.68 - $590.89

Specifications

Color Clear, colorless
Concentration 0.00500N ±0.00005N (0.0025M)
pH Range 6.0 to 7.0
Format Liquid
Packaging Amber Glass
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
LC183402
View Documents Promotion Details
N/A 1 L
Each for $193.68
Only null left
 
LC183404
View Documents Promotion Details
N/A 4 L
Each for $590.89
Only null left
 
Description

Description

Specifications

Specifications

Clear, colorless
6.0 to 7.0
Amber Glass
Phenylarsine Oxide
0.00500N ±0.00005N (0.0025M)
Liquid
Room Temperature
Videos
SDS
Documents

Documents