missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylarsine Oxide, For Dissolved Oxygen, Certified, 0.00500N ±0.00005N (0.0025M), LabChem™
Specifications
Specifications
| Color | Clear, colorless |
| Concentration | 0.00500N ±0.00005N (0.0025M) |
| pH Range | 6.0 to 7.0 |
| Format | Liquid |
| Packaging | Amber Glass |
| Quantity | 4 L |
| Storage Requirements | Room Temperature |
| Type | Phenylarsine Oxide |