missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide, Certified, 0.025N ±0.001N (0.0125M), LabChem™

Catalog No. p-5388328
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
500 mL
Packaging:
Amber Glass
Catalog No. Quantity Packaging
LC183801 500 mL Amber Glass
LC183802 1 L Amber Glass
2 options

Catalog No. LC183801

Supplier: LabChem LC183801

Only null left

Chemical Identifiers

CAS 637-03-6 , 7732-18-5
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

CAS Max % 99.9, 0.1
Color Colorless
pH 6.0 to 7.0
Packaging Amber Glass
Linear Formula C6H5AsO
Quantity 500 mL
Solubility Information Soluble in water
Formula Weight 168.03
Grade Certified
Concentration or Composition (by Analyte or Components) 0.025N ±0.001N (0.0125M)
Physical Form Liquid
Chemical Name or Material Phenylarsine Oxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature