missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide, (1mL = 0.2mg Cl), Certified, 0.00564N ±0.00005N (0.00282M), LabChem™

Catalog No. LC183604
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC183604 4 L Amber Glass
LC183601 500 mL Amber Glass
LC183602 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC183604 Supplier LabChem Supplier No. LC183604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-03-6 , 7732-18-5
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

CAS Max % 99.9, 0.1
Color Colorless
pH 6.0 to 7.0
Packaging Amber Glass
Linear Formula C6H5AsO
Quantity 4 L
Solubility Information Soluble in water
Formula Weight 168.03
Grade Certified
Concentration or Composition (by Analyte or Components) 0.00564N ±0.00005N (0.00282M)
Physical Form Liquid
Chemical Name or Material Phenylarsine Oxide, (1mL = 0.2mg Cl)
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature