missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylacetic Hydrazide 98.0+%, TCI America™
Supplier: TCI America P220325G
Specifications
| Phenylacetic Hydrazide | |
| 117°C | |
| MFCD00007612 | |
| phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide | |
| C1=CC=C(C=C1)CC(=O)NN | |
| 150.181 | |
| 150.18 | |
| Crystalline Powder |
| 937-39-3 | |
| C8H10N2O | |
| 25 g | |
| FPTCVTJCJMVIDV-UHFFFAOYSA-N | |
| 2-phenylacetohydrazide | |
| 70301 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 937-39-3 | |
| 150.181 | |
| FPTCVTJCJMVIDV-UHFFFAOYSA-N | |
| 70301 | |
| C1=CC=C(C=C1)CC(=O)NN |
| C8H10N2O | |
| MFCD00007612 | |
| phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide | |
| 2-phenylacetohydrazide |
Safety and Handling
TSCA : Yes