missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Vinyl Sulfone 98.0+%, TCI America™

Catalog No. p-7134885
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
P09825G 5 g
P098225G 25 g
2 options

Catalog No. P09825G

Supplier: TCI America P09825G

Only null left

Chemical Identifiers

CAS 5535-48-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00007554
InChI Key UJTPZISIAWDGFF-UHFFFAOYSA-N
Synonym phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon
PubChem CID 79664
IUPAC Name (ethenesulfonyl)benzene
SMILES C=CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Vinyl Sulfone
EINECSNumber (3)-4398
TSCA No