missing translation for 'onlineSavingsMsg'
Learn More

Phenyl vinyl sulfone, 98%

Catalog No. AC221250010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC221250010 1 g
AC221250050 5 g
AC221250250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221250010 Supplier Thermo Scientific Chemicals Supplier No. 221250010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5535-48-8 | C8H8O2S | 168.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5535-48-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00007554
InChI Key UJTPZISIAWDGFF-UHFFFAOYSA-N
Synonym phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon
PubChem CID 79664
IUPAC Name ethenylsulfonylbenzene
SMILES C=CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 66.0°C to 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SO2CH=CH2
Quantity 1 g
Fieser 10,315; 13,335
Merck Index 15,7431
Solubility Information Solubility in water: slightly soluble.
Formula Weight 168.21
Percent Purity 98%
Chemical Name or Material Phenyl vinyl sulfone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
EINECSNumber 226-890-2

RUO – Research Use Only