missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. P18291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18291G 1 g
P18295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18291G Supplier TCI America Supplier No. P18291G

Chemical Identifiers

CAS 17763-67-6
Molecular Formula C7H5F3O3S
Molecular Weight (g/mol) 226.169
MDL Number MFCD00192399
InChI Key GRJHONXDTNBDTC-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid Phenyl Ester, Phenyl Triflate
PubChem CID 548641
IUPAC Name phenyl trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 62°C
Quantity 1 g
Formula Weight 226.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Trifluoromethanesulfonate