missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Phenyl Sepharose™ High Performance Media

Catalog No. 45002946
Change view
Click to view available options
Quantity:
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002946 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002946 Supplier Cytiva Supplier No. 17108201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for intermediate and polishing step purification steps

  • Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked agarose beads with aromatic phenyl functional groups
  • High resolution due to smaller 34μm bead size
  • Chemical stability for effective CIP and sanitization
  • Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes

Specifications

Quantity 75 mL
For Use With (Application) For Intermediate PurifiCatIon and Polishing AppliCatIons
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 1 M Acetic Acid, 30% Acetonitrile, 70% Ethanol, 30% Isopropanol, 6 M Guanidine-Hydrochloride, 8 M Urea, , 3 M Ammonium sulfate, Ionic Detergents, Non-ionic detergents, polar organic solvents, 2% Sodium Dodectyl Sulfate
Storage Requirements 4°C to 30°C
Product Type Phenyl Sepharose High Performance Media
Flow Rate 100 to 200 cm/Hoμr
Particle Size 24 to 44 μm
Height (English) Bed 11.8 in.
Height (Metric) Bed 30 cm
Max. Pressure 300 kPa
Matrix 6% Cross-Linked Agarose
pH Range 3 to 13
Storage Buffer 20% Ethanol
Ligand Type Phenyl
Show More Show Less