missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Salicylate 98.0+%, TCI America™

Catalog No. S001725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001725G 25 g
S0017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001725G Supplier TCI America Supplier No. S001725G

Chemical Identifiers

CAS 118-55-8
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002213
InChI Key ZQBAKBUEJOMQEX-UHFFFAOYSA-N
Synonym phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
PubChem CID 8361
ChEBI CHEBI:34918
IUPAC Name phenyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O

Specifications

Melting Point 44°C
Color White
Boiling Point 173°C
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Salicylate
EINECSNumber (3)-1597
RTECSNumber VO6125000
TSCA Yes