missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Salicylate, Crystal, Purified, 98%, Spectrum™ Chemical

Catalog No. p-7166211
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
40 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612283 125 g Poly Bottle
18612284 500 g Poly Bottle
18612285 2.5 kg Poly Bottle
18612286 12 kg Poly Pail
18612287 40 kg Fiber Drum
18612288 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18612283 Supplier Spectrum Chemical Mfg Cor Supplier No. PH160125GM
Only null left

Chemical Identifiers

CAS 118-55-8

Specifications

CAS Max % 1
Melting Point 40° to 47°C
Packaging Poly Bottle
Quantity 125 g
Percent Purity 0.98
Grade Purified

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.