missing translation for 'onlineSavingsMsg'
Learn More

Phenyl salicylate, 99%

Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC) | CAS: 118-55-8 | C13H10O3 | 214.22 g/mol

Supplier:  Thermo Scientific Chemicals 165080010

 View more versions of this product

Catalog No. AC165080010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenyl salicylate
40.0°C to 44.0°C
172.0°C to 173.0°C (12.0 mmHg)
Authentic
Glass bottle
2-(HO)C6H4CO2C6H5
10,76
phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
ZQBAKBUEJOMQEX-UHFFFAOYSA-N
phenyl 2-hydroxybenzoate
8361
214.22
Fine Crystalline Powder
118-55-8
White
>110°C
98.5% min. (GC)
C13H10O3
1 kg
15,742
Solubility in water: <0.1% (25°C). Other solubilities: 1g/1.5mL benzene- 1g/5mL amyl alcohol,53g/100g abs.ethanol-470g/100g ethylacetate (25 c),620g/100g methylethylketone-460g/100g toluene(25 c,soluble in chloroform,acetone,ether and oils,s
C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O
214.22
CHEBI:34918
99%

Chemical Identifiers

118-55-8
214.22
phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
CHEBI:34918
C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O
C13H10O3
ZQBAKBUEJOMQEX-UHFFFAOYSA-N
8361
phenyl 2-hydroxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 204-259-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only