missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Propargyl Ether 98.0+%, TCI America™
Supplier: TCI America P222225G
Specifications
| Phenyl Propargyl Ether | |
| Yellow | |
| C9H8O | |
| 25 g | |
| AIQRJSXKXVZCJO-UHFFFAOYSA-N | |
| (prop-2-yn-1-yloxy)benzene | |
| 83613 | |
| ≥98.0% (GC) |
| 13610-02-1 | |
| 83°C | |
| MFCD00048107 | |
| phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl | |
| C#CCOC1=CC=CC=C1 | |
| 132.16 | |
| 132.16 | |
| Liquid |
Chemical Identifiers
| 13610-02-1 | |
| 132.16 | |
| AIQRJSXKXVZCJO-UHFFFAOYSA-N | |
| 83613 | |
| C#CCOC1=CC=CC=C1 |
| C9H8O | |
| MFCD00048107 | |
| phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl | |
| (prop-2-yn-1-yloxy)benzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator