missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Propargyl Ether 98.0+%, TCI America™

Catalog No. p-7135464
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P22225G 5 g
P222225G 25 g
2 options

Catalog No. P22225G

Supplier: TCI America P22225G

Only null left

Chemical Identifiers

CAS 13610-02-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00048107
InChI Key AIQRJSXKXVZCJO-UHFFFAOYSA-N
Synonym phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
PubChem CID 83613
IUPAC Name (prop-2-yn-1-yloxy)benzene
SMILES C#CCOC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Propargyl Ether
TSCA No
Recommended Storage Refrigerator