missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphorodichloridate, 97%

Catalog No. aaa1047922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1047922 100 g
AAA1047914 25 g
AAA1047936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1047922 Supplier Thermo Scientific Chemicals Supplier No. A1047922
Only null left

CAS: 770-12-7 | C6H5Cl2O2P | 210.978 g/mol

Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, alcohols and strong oxidizing agents.

Chemical Identifiers

CAS 770-12-7
Molecular Formula C6H5Cl2O2P
Molecular Weight (g/mol) 210.978
MDL Number MFCD00002067
InChI Key TXFOLHZMICYNRM-UHFFFAOYSA-N
Synonym phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester
PubChem CID 13038
IUPAC Name dichlorophosphoryloxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(Cl)Cl

Specifications

Melting Point -1°C
Density 1.415
Boiling Point 241°C to 243°C
Flash Point 112°C (233°F)
Refractive Index 1.523
Quantity 100 g
UN Number UN3265
Beilstein 511863
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 210.98
Percent Purity 97%
Chemical Name or Material Phenyl phosphorodichloridate
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-220-6
RTECSNumber TD4393000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only