missing translation for 'onlineSavingsMsg'
Learn More

Phenyl p-tolyl sulfone, 98+%

Catalog No. AAL0312809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0312809 10 g
AAL0312818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0312809 Supplier Thermo Scientific Chemicals Supplier No. L0312809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 640-57-3 | C13H12O2S | 232.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 640-57-3
Molecular Formula C13H12O2S
Molecular Weight (g/mol) 232.297
MDL Number MFCD00025038
InChI Key YBRXHRWLEFCFEG-UHFFFAOYSA-N
Synonym phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene
PubChem CID 69497
IUPAC Name 1-(benzenesulfonyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 125°C to 129°C
Odor Odorless
Quantity 10 g
Beilstein 1872872
Formula Weight 232.3
Percent Purity ≥98%
Chemical Name or Material Phenyl p-tolyl sulfone
EINECSNumber 211-364-7
RTECSNumber WR6880000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only