missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Methacrylate (stabilized with BHT) 97.0+%, TCI America™
Supplier: TCI America P213425G
Specifications
| Phenyl Methacrylate (stabilized with BHT) | |
| Yellow | |
| C10H10O2 | |
| 25 g | |
| phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht | |
| CC(=C)C(=O)OC1=CC=CC=C1 | |
| 162.188 | |
| 162.19 | |
| Liquid |
| 2177-70-0 | |
| 198°C | |
| MFCD00048117 | |
| 3272 | |
| QIWKUEJZZCOPFV-UHFFFAOYSA-N | |
| phenyl 2-methylprop-2-enoate | |
| 75121 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 2177-70-0 | |
| 162.188 | |
| QIWKUEJZZCOPFV-UHFFFAOYSA-N | |
| 75121 | |
| CC(=C)C(=O)OC1=CC=CC=C1 |
| C10H10O2 | |
| MFCD00048117 | |
| phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht | |
| phenyl 2-methylprop-2-enoate |
Safety and Handling
EINECSNumber : (3)-4128
TSCA : Yes