missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Methacrylate (stabilized with BHT) 97.0+%, TCI America™

Catalog No. P21345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21345G 5 g
P213425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21345G Supplier TCI America Supplier No. P21345G
Only null left

Chemical Identifiers

CAS 2177-70-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00048117
InChI Key QIWKUEJZZCOPFV-UHFFFAOYSA-N
Synonym phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht
PubChem CID 75121
IUPAC Name phenyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 5 g
UN Number 3272
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Methacrylate (stabilized with BHT)
EINECSNumber (3)-4128
TSCA Yes