missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Dichlorophosphate 98.0+%, TCI America™

Catalog No. p-7134489
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020925G 25 g
P0209500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P020925G Supplier TCI America Supplier No. P020925G
Only null left

Chemical Identifiers

CAS 770-12-7
Molecular Formula C6H5Cl2O2P
Molecular Weight (g/mol) 210.978
MDL Number MFCD00002067
InChI Key TXFOLHZMICYNRM-UHFFFAOYSA-N
Synonym phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester
PubChem CID 13038
IUPAC Name dichlorophosphoryloxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(Cl)Cl

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
UN Number 3265
Formula Weight 210.98
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Phenyl Dichlorophosphate
EINECSNumber (3)-4457
RTECSNumber TD4393000
TSCA Yes