missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Chloroformate 98.0+%, TCI America™
$38.51 - $206.00
Chemical Identifiers
| CAS | 1885-14-9 |
|---|---|
| Molecular Formula | C7H5ClO2 |
| Molecular Weight (g/mol) | 156.57 |
| MDL Number | MFCD00000637 |
| InChI Key | AHWALFGBDFAJAI-UHFFFAOYSA-N |
| Synonym | phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y |
| PubChem CID | 15891 |
| IUPAC Name | phenyl carbonochloridate |
| SMILES | ClC(=O)OC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
C064925ML
|
TCI America
C064925ML |
25 mL |
Each for $38.51
|
|
|||||
|
C0649100ML
|
TCI America
C0649100ML |
100 mL |
Each for $65.46
|
|
|||||
|
C0649500ML
|
TCI America
C0649500ML |
500 mL |
Each for $206.00
|
|
|||||
Chemical Identifiers
| 1885-14-9 | |
| 156.57 | |
| AHWALFGBDFAJAI-UHFFFAOYSA-N | |
| 15891 | |
| ClC(=O)OC1=CC=CC=C1 |
| C7H5ClO2 | |
| MFCD00000637 | |
| phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
| phenyl carbonochloridate |
Specifications
| 1885-14-9 | |
| 187°C | |
| MFCD00000637 | |
| 2746 | |
| AHWALFGBDFAJAI-UHFFFAOYSA-N | |
| phenyl carbonochloridate | |
| 15891 | |
| ≥98.0% (GC,T) | |
| Phenyl Chloroformate |
| Yellow | |
| C7H5ClO2 | |
| 25 mL | |
| phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
| ClC(=O)OC1=CC=CC=C1 | |
| 156.57 | |
| 156.57 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0629
RTECSNumber : FG3850000
TSCA : Yes
Recommended Storage : Refrigerator