missing translation for 'onlineSavingsMsg'
Learn More

Phenyl chloroformate, 99%

CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol

$68.33 - $247.14

Chemical Identifiers

CAS 1885-14-9
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00000637
InChI Key AHWALFGBDFAJAI-UHFFFAOYSA-N
Synonym phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
PubChem CID 15891
IUPAC Name phenyl carbonochloridate
SMILES ClC(=O)OC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1261822
View Documents
Thermo Scientific Chemicals
A1261822
100 g N/A
Only null left
 
AAA1261830
View Documents
Thermo Scientific Chemicals
A1261830
250 g N/A
Only null left
 
AAA1261836
View Documents
Thermo Scientific Chemicals
A1261836
500 g N/A
Only null left
 
Description

Description

Phenyl chloroformate is used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate. It acts as a precursor of phenyl mixed anhydrides which are used in peptide coupling reactions. It serves as a dehydrating reagent for the conversion of primary amides to nitriles and an intermediate in the synthesis of pharmaceuticals and carbamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl chloroformate is used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate. It acts as a precursor of phenyl mixed anhydrides which are used in peptide coupling reactions. It serves as a dehydrating reagent for the conversion of primary amides to nitriles and an intermediate in the synthesis of pharmaceuticals and carbamates.

Solubility
Miscible with N,N-dimethylformamide.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, alcohols and amines. Hydrolyses in water.
Specifications

Chemical Identifiers

1885-14-9
156.57
AHWALFGBDFAJAI-UHFFFAOYSA-N
15891
ClC(=O)OC1=CC=CC=C1
C7H5ClO2
MFCD00000637
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
phenyl carbonochloridate

Specifications

1885-14-9
1.246
76°C (169°F)
1.511
100 g
606778
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
AHWALFGBDFAJAI-UHFFFAOYSA-N
phenyl carbonochloridate
15891
99%
-38°C
188°C to 189°C
C7H5ClO2
MFCD00000637
UN2746
Moisture sensitive
Miscible with N,N-dimethylformamide.
ClC(=O)OC1=CC=CC=C1
156.57
156.57
Phenyl chloroformate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318-H227-H302-H317
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
May cause an allergic skin reaction.

P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H227-H302-H314-H317-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENYL CHLOROFORMATE

EINECSNumber : 217-547-8

RTECSNumber : FG3850000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only