missing translation for 'onlineSavingsMsg'
Learn More

Phenyl carbamate, 98+%

Catalog No. p-7043478
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0094406 5 g
AAL0094414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0094406 Supplier Thermo Scientific Chemicals Supplier No. L0094406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 622-46-8 | C7H7NO2 | 137.138 g/mol

Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 622-46-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007961
InChI Key BSCCSDNZEIHXOK-UHFFFAOYSA-N
Synonym carbamic acid, phenyl ester, phenol carbamate, carbamic acid phenyl ester, unii-jkb257u27v, o-phenyl carbamate, ccris 5071, phenyl aminooate, mono-phenylcarbamate, mono-phenol carbamate, phenyl carbamate
PubChem CID 69322
IUPAC Name phenyl carbamate
SMILES C1=CC=C(C=C1)OC(=O)N

Specifications

Melting Point 149°C to 152°C
Quantity 5 g
Beilstein 2042627
Solubility Information Soluble in water.
Formula Weight 137.14
Percent Purity ≥98%
Chemical Name or Material Phenyl carbamate
EINECSNumber 210-737-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only