missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Bromoacetate 95.0+%, TCI America™

Catalog No. P193625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P193625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P193625G Supplier TCI America Supplier No. P193625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 620-72-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00192391
InChI Key UEWYUCGVQMZMGY-UHFFFAOYSA-N
Synonym phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
PubChem CID 564919
IUPAC Name phenyl 2-bromoacetate
SMILES C1=CC=C(C=C1)OC(=O)CBr

Specifications

Boiling Point 112°C
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Phenyl Bromoacetate