missing translation for 'onlineSavingsMsg'
Learn More

Phenyl bromoacetate, 98%

Catalog No. AAH6046406
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6046406 5 g
AAH6046418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6046406 Supplier Thermo Scientific Chemicals Supplier No. H6046406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 620-72-4 | C8H7BrO2 | 215.046 g/mol

Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

Solubility
Soluble in ethanol and ether, insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.

Chemical Identifiers

CAS 620-72-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00192391
InChI Key UEWYUCGVQMZMGY-UHFFFAOYSA-N
Synonym phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
PubChem CID 564919
IUPAC Name phenyl 2-bromoacetate
SMILES C1=CC=C(C=C1)OC(=O)CBr

Specifications

Melting Point 31°C to 33°C
Density 1.508
Boiling Point 134°C
Flash Point 113°C (235°F)
Linear Formula BrCH2CO2C6H5
Quantity 5 g
Solubility Information Soluble in ethanol and ether,insoluble in water.
Formula Weight 215.04
Percent Purity 98%
Chemical Name or Material Phenyl bromoacetate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only