missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Acetate 98.0+%, TCI America™

Catalog No. A004325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A004325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A004325G Supplier TCI America Supplier No. A004325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-79-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00008699
InChI Key IPBVNPXQWQGGJP-UHFFFAOYSA-N
Synonym acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech
PubChem CID 31229
ChEBI CHEBI:8082
IUPAC Name phenyl acetate
SMILES CC(=O)OC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 195°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Acetate
EINECSNumber (3)-0643
RTECSNumber AJ2800000
TSCA Yes