missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 4,6-O-Benzylidene-1-thio-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. p14755g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14755G Supplier TCI America Supplier No. P14755G
Only null left

Chemical Identifiers

CAS 87508-17-6
Molecular Formula C19H20O5S
Molecular Weight (g/mol) 360.424
MDL Number MFCD06797160
InChI Key BDNIQCYVYFGHSI-YETGYGHFSA-N
PubChem CID 133556307
IUPAC Name (4aS,6R,7S,8S,8aR)-2-phenyl-6-phenylsulfanyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
SMILES C1C2C(C(C(C(O2)SC3=CC=CC=C3)O)O)OC(O1)C4=CC=CC=C4

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 360.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 4,6-O-Benzylidene-1-thio-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer