missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 3-Chloropropionate 95.0+%, TCI America™

Catalog No. C085025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C085025ML 25 mL
C0850500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C085025ML Supplier TCI America Supplier No. C085025ML

Chemical Identifiers

CAS 24552-27-0
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00045298
InChI Key RAFRTSDUWORDLA-UHFFFAOYSA-N
Synonym 3-Chloropropionic Acid Phenyl Ester
PubChem CID 141118
IUPAC Name phenyl 3-chloropropanoate
SMILES C1=CC=C(C=C1)OC(=O)CCCl

Specifications

Color Yellow
Boiling Point 157°C
Quantity 25 mL
Formula Weight 184.62
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl 3-Chloropropionate