missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl[2-(trimethylsilyl)phenyl]iodonium Trifluoromethanesulfonate 97.0+%, TCI America™
Supplier: TCI America P16205G
Specifications
| Phenyl[2-(trimethylsilyl)phenyl]iodonium Trifluoromethanesulfonate | |
| 132°C | |
| C16H18F3IO3SSi | |
| Phenyl[2-(trimethylsilyl)phenyl]iodonium Triflate | |
| C[Si](C)(C)C1=CC=CC=C1[I+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-] | |
| 502.362 | |
| 502.36 | |
| Crystalline Powder |
| 164594-13-2 | |
| White | |
| 5 g | |
| GSCTXOAWWNOUFK-UHFFFAOYSA-M | |
| phenyl-(2-trimethylsilylphenyl)iodanium;trifluoromethanesulfonate | |
| 10885617 | |
| ≥97.0% (HPLC,T) |
Chemical Identifiers
| 164594-13-2 | |
| 502.362 | |
| Phenyl[2-(trimethylsilyl)phenyl]iodonium Triflate | |
| phenyl-(2-trimethylsilylphenyl)iodanium;trifluoromethanesulfonate |
| C16H18F3IO3SSi | |
| GSCTXOAWWNOUFK-UHFFFAOYSA-M | |
| 10885617 | |
| C[Si](C)(C)C1=CC=CC=C1[I+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-] |
Safety and Handling
TSCA : No