missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2-Pyridyl Ketoxime 98.0+%, TCI America™

Catalog No. P02235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02235G 5 g
P02231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02235G Supplier TCI America Supplier No. P02235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1826-28-4
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00006310
InChI Key WBSIGZZXFQOEJK-QXMHVHEDSA-N
Synonym phenylpyridylketoxime, phenyl pyridin-2-yl methanone oxime, phenyl 2-pyridyl ketone oxime, methanone, phenyl-2-pyridinyl-, oxime, 2-benzoylpyridiine ketoxime, phenyl 2-pyridyl methanone oxime, 2-benzoylpyridine oxime, e-phenyl-2-pyridyl ketone oxime, hydroxyimino phenyl-2-pyridylmethane, ketone, phenyl 2-pyridyl, oxime 8ci
PubChem CID 5397197
IUPAC Name (2Z)-2-[nitroso(phenyl)methylidene]-1,2-dihydropyridine
SMILES O=N\C(=C1/NC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 152°C
Color White
Quantity 5 g
Formula Weight 198.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 2-Pyridyl Ketoxime